4-Propoxy-3-methylphenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Methyl-4-propoxybenzeneboronic acid, (3-Methyl-4-propoxyphenyl)boronic acid

A326251-1G A326251-250MG
76693-858EA 60 USD
76693-858 76693-860
4-Propoxy-3-methylphenylboronic acid 97%
4-Propoxy-3-methylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MDL Number: MFCD06659872
CAS Number: 279262-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR