Bilastine 98%

Supplier: Ambeed
Warning

Synonyms: 2-(4-(2-(4-(1-(2-Ethoxyethyl)-1H-benzo[d]imidazol-2-yl)piperidin-1-yl)ethyl)phenyl)-2-methylpropanoic acid, 2-[4-[2-[4-[1-(2-Ethoxyethyl)-1H-benzo[d]imidazol-2-yl]piperidin-1-yl]ethyl]phenyl]-2-methylpropanoic acid

A325945-100MG A325945-10MG A325945-1G A325945-1MG A325945-50MG A325945-5MG A325945-250MG
76693-846EA 40 USD
76693-846 76693-848 76693-850 76693-852 76693-854 76693-856 77626-610
Bilastine 98%
Bilastine

Precaución: for Research Use Only

Formula: C₂₈H₃₇N₃O₃
MW: 463.62 g/mol
MDL Number: MFCD09837814
CAS Number: 202189-78-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR