2,5-Xylylacetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,5-Dimethylphenyl)acetic acid, (2,5-Dimethylphenyl)acetic acid

A325355-100G A325355-10G A325355-1G A325355-25G A325355-5G
76693-834EA 91.24 USD
76693-834 76693-836 76693-838 76693-840 76693-842
2,5-Xylylacetic acid 98%
2,5-Xylylacetic acid

Precaución: for Research Use Only

Formula: (C₆H₃(CH₃)₂)CH₂COOH
MW: 164.2 g/mol
MDL Number: MFCD00014369
CAS Number: 13612-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR