Sitagliptin phosphate monohydrate 99%

Supplier: Ambeed
Warning

Synonyms: (R)-3-Amino-1-(3-(trifluoromethyl)-5,6-dihydro-[1,2,4]triazolo[4,3-a]pyrazin-7(8H)-yl)-4-(2,4,5-trifluorophenyl)butan-1-one phosphate hydrate

A323507-10G A323507-1G A323507-250MG A323507-5G A323507-100MG A323507-100G A323507-25G
76693-816EA 40 USD
76693-816 76693-818 76693-820 76693-822 77593-920 77626-596 77626-598
Sitagliptin phosphate monohydrate 99%
Sitagliptin phosphate monohydrate

Precaución: for Research Use Only

Formula: C₁₆H₂₀F₆N₅O₆P
MW: 523.32 g/mol
MDL Number: MFCD19684081
CAS Number: 654671-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR