Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer 98%

Supplier: Ambeed
Warning

A323351-100MG A323351-250MG
76693-810EA 74.27 USD
76693-810 76693-812
Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer 98%
Chloro(pentamethylcyclopentadienyl)ruthenium(II) tetramer

Precaución: for Research Use Only

Formula: C₄₀H₆₀Ru₄Cl₄
MW: 1087.0000 g/mol
MDL Number: MFCD03792150
CAS Number: 113860-07-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR