cis-1,2-Cyclopentanedicarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: cis-Cyclopentane-1,2-dicarboxylic acid

A322752-1G A322752-5G
76693-802EA 82.76 USD
76693-802 76693-804
cis-1,2-Cyclopentanedicarboxylic acid 97%
cis-1,2-Cyclopentanedicarboxylic acid

Precaución: for Research Use Only

Formula: C₇H₁₀O₄
MW: 158.16 g/mol
MDL Number: MFCD01311172
CAS Number: 1461-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR