Calix[6]arene 98%

Supplier: Ambeed
Danger

A321030-1G A321030-250MG A321030-25G
76693-782EA 63.66 USD
76693-782 76693-784 76693-786
Calix[6]arene 98%
Calix[6]arene

Precaución: for Research Use Only

Formula: C₄₂H₃₆O₆
MW: 636.74 g/mol
MDL Number: MFCD00143083
CAS Number: 96107-95-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR