Benzyl-D-(-)-mandelate 98%

Supplier: Ambeed
Warning

Synonyms: D-(-)-Mandelic Acid Benzyl Ester, (R)-Benzyl 2-hydroxy-2-phenylacetate

A320991-100G A320991-10G A320991-5G
76693-776EA 42.44 USD
76693-776 76693-778 76693-780
Benzyl-D-(-)-mandelate 98%
Benzyl-D-(-)-mandelate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₃
MW: 242.27 g/mol
MDL Number: MFCD00674031
CAS Number: 97415-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR