(E)-5-(2-Bromovinyl)-2'-deoxyuridine 95%

Supplier: Ambeed
Warning

A320199-1MG A320199-250MG A320199-25MG A320199-50MG
76693-770EA 5 USD
76693-770 76693-772 76693-774 77878-013
(E)-5-(2-Bromovinyl)-2'-deoxyuridine 95%
(E)-5-(2-Bromovinyl)-2'-deoxyuridine

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrN₂O₅
MDL Number: MFCD00058585
CAS Number: 69304-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR