Bis(aminoguanidinium) sulfate 98%

Supplier: Ambeed
Warning

A319868-1000G A319868-100G A319868-25G A319868-500G
76693-750EA 253 USD
76693-750 76693-752 76693-754 76693-756
Bis(aminoguanidinium) sulfate 98%
Bis(aminoguanidinium)sulphate

Precaución: for Research Use Only

Formula: C₂H₁₄N₈O₄S
MDL Number: MFCD00035665
CAS Number: 996-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR