Cyanuric acid 98%

Supplier: Ambeed
Warning

A318404-1000G A318404-100G A318404-25G A318404-500G
76693-732EA 19 USD
76693-732 76693-734 76693-736 76693-738
Cyanuric acid 98%
Cyanuric acid

Precaución: for Research Use Only

Formula: C₃H₃N₃O₃
MDL Number: MFCD00082990
CAS Number: 108-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR