Diallyl diisopropylphosphoramidite 95%

Supplier: Ambeed
Danger

A318264-1G A318264-250MG
76693-728EA 108.22 USD
76693-728 76693-730
Diallyl diisopropylphosphoramidite 95%
Diallyl diisopropylphosphoramidite

Precaución: for Research Use Only

Formula: C₁₂H₂₄NO₂P
MW: 245.29 g/mol
MDL Number: MFCD00216647
CAS Number: 126429-21-8
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR