Boc-DL-3-aminobutanoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-((tert-Butoxycarbonyl)amino)butanoic acid

A317388-1G A317388-250MG A317388-25G A317388-5G
76693-718EA 8 USD
76693-718 76693-720 76693-722 76693-724
Boc-DL-3-aminobutanoic acid 97%
Boc-DL-3-aminobutanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MDL Number: MFCD00270338
CAS Number: 52815-19-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR