Sorafenib (BAY 43-9006) 99%

Supplier: Ambeed
Danger

A316727-100G A316727-1G A316727-25G A316727-5G A316727-50MG A316727-10MG
76693-704EA 287 USD
76693-704 76693-706 76693-708 76693-710 77601-424 77626-578
Sorafenib (BAY 43-9006) 99%
Sorafenib (BAY 43-9006)

Precaución: for Research Use Only

Formula: C₂₁H₁₆ClF₃N₄O₃
MDL Number: MFCD06411450
CAS Number: 284461-73-0
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR