Clofazimine 98%

Supplier: Ambeed
Warning

Synonyms: N,5-Bis(4-chlorophenyl)-3,5-dihydro-3-(isopropylimino)phenazin-2-amine, N,5-Bis(4-chlorophenyl)-3-(isopropylimino)-3,5-dihydrophenazin-2-amine

A316630-100MG A316630-1G A316630-1MG A316630-250MG A316630-25G A316630-5G
76693-692EA 7.43 USD
76693-692 76693-694 76693-696 76693-698 76693-700 76693-702
Clofazimine 98%
Clofazimine

Precaución: for Research Use Only

Formula: C₂₇H₂₂Cl₂N₄
MW: 473.40 g/mol
MDL Number: MFCD00056793
CAS Number: 2030-63-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR