2-Ethoxy-5-(trifluoromethyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 2-Ethoxy-5-trifluoromethylphenylboronic acid

A315914-1G A315914-250MG
76693-680EA 37.14 USD
76693-680 76693-682
2-Ethoxy-5-(trifluoromethyl)phenylboronic acid 98%
2-Ethoxy-5-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BF₃O₃
MW: 233.98 g/mol
MDL Number: MFCD06801695
CAS Number: 850593-10-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR