Methyl 2-naphthylacetate 97%

Supplier: Ambeed
Warning

A314345-100MG A314345-1G A314345-250MG A314345-5G
76693-654EA 42 USD
76693-654 76693-656 76693-658 76693-660
Methyl 2-naphthylacetate 97%
Methyl 2-naphthylacetate

Precaución: for Research Use Only

Formula: C₁₃H₁₂O₂
MDL Number: MFCD09263745
CAS Number: 2876-71-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR