(2S,5R)-Benzhydryl 3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide 95%

Supplier: Ambeed
Warning

A313841-100G A313841-1G A313841-250MG A313841-25G A313841-5G
76693-642EA 872.06 USD
76693-642 76693-644 76693-646 76693-648 76693-650
(2S,5R)-Benzhydryl 3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide 95%
(2S,5R)-Benzhydryl 3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate 4-oxide

Precaución: for Research Use Only

Formula: C₂₁H₂₁NO₄S
MW: 383.46 g/mol
MDL Number: MFCD11044527
CAS Number: 87579-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR