(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97%, ee 99%
Supplier: Ambeed
|
Warning
|
A313231-100MG
A313231-1G
A313231-250MG
A313231-5G
76693-628EA
10.61
USD
76693-628
76693-630
76693-632
76693-634
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol ≥97%, ee 99%
(R)-(+)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol
Precaución: for Research Use Only
Formula:
C₂₀H₂₀Br₂O₂ |
MDL Number:
MFCD03093977 CAS Number: 65355-08-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...