Amezinium methyl sulfate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulfate, Amezinium Methyl sulphate, Lu-1631, 4-Amino-6-methoxy-1-phenylpyridazinium Methyl Sulphate, Amezinium methylsulphate, Amezinium metilsulfate, 4-Amino-6-methoxy-1-phenylpyridazin-1-ium methyl sulphate

A313059-100MG A313059-1MG A313059-25MG
76693-616EA 8.49 USD
76693-616 76693-618 76693-620
Amezinium methyl sulfate 98%
Amezinium methyl sulphate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₅S
MW: 313.33 g/mol
MDL Number: MFCD00865972
CAS Number: 30578-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR