(R)-3-((tert-Butoxycarbonyl)amino)-3-(4-hydroxyphenyl)propanoic acid 98+%

Supplier: Ambeed
Warning

A312657-100MG A312657-1G A312657-250MG
76693-604EA 54.11 USD
76693-604 76693-606 76693-608
(R)-3-((tert-Butoxycarbonyl)amino)-3-(4-hydroxyphenyl)propanoic acid 98+%
(R)-3-((tert-Butoxycarbonyl)amino)-3-(4-hydroxyphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₅
MW: 281.30 g/mol
MDL Number: MFCD03427941
CAS Number: 329013-12-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR