Bis(2-methoxyethoxy)methane 97%

Supplier: Ambeed

A311593-100G A311593-25G A311593-500G
76693-594EA 66 USD
76693-594 76693-596 77626-554
Bis(2-methoxyethoxy)methane 97%
Bis(2-methoxyethoxy)methane

Precaución: for Research Use Only

Formula: C₇H₁₆O₄
MDL Number: MFCD00043945
CAS Number: 4431-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR