Alsterpaullone 98%

Supplier: Ambeed
Warning

Synonyms: 7,12-Dihydro-9-nitroindolo[3,2-d][1]benzazepin-6(5H)-one

A310955-1MG A310955-5MG
76693-588EA 20 USD
76693-588 76693-590
Alsterpaullone 98%
Alsterpaullone

Precaución: for Research Use Only

Formula: C₁₆H₁₁N₃O₃
MDL Number: MFCD02683579
CAS Number: 237430-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR