3,5-Xylyl isothiocyanate 95%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dimethylphenyl isothiocyanate, 3,5-Dimethylphenylisothiocyanate

A310849-1G A310849-25G A310849-5G
76693-582EA 28.65 USD
76693-582 76693-584 76693-586
3,5-Xylyl isothiocyanate 95%
3,5-Xylyl isothiocyanate

Precaución: for Research Use Only

Formula: C₉H₉NS
MW: 163.24 g/mol
MDL Number: MFCD00041082
CAS Number: 40046-30-8
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR