4-Chloro-2-methylanisole 95%

Supplier: Ambeed
Warning

A310751-5G A310751-25G
76693-580EA 20 USD
76693-580 77877-989
4-Chloro-2-methylanisole 95%
4-Chloro-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉ClO
MDL Number: MFCD00070771
CAS Number: 3260-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR