Abacavir 98%

Supplier: Ambeed
Danger

Synonyms: (1S,4R)-4-[2-Amino-6-(cyclopropylamino)-9H-purin-9-yl]-2-cyclopentene-1-methanol, ((1S,4R)-4-(2-Amino-6-(cyclopropylamino)-9H-purin-9-yl)cyclopent-2-en-1-yl)methanol

A310646-10MG A310646-1G A310646-25G A310646-50MG A310646-5G A310646-100G
76693-562EA 5.31 USD
76693-562 76693-564 76693-566 76693-568 76693-570 77601-404
Abacavir 98%
Abacavir

Precaución: for Research Use Only

Formula: C₁₄H₁₈N₆O
MW: 286.34 g/mol
MDL Number: MFCD00903850
CAS Number: 136470-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR