ɑ-Methylene-γ-butyrolactone 95% stabilized

Supplier: Ambeed
Danger


Stabilization: Stabilised with 2,6-Di-tert-butyl-p-cresol 2%

A309252-1G A309252-25G A309252-5G
76693-538EA 12.74 USD
76693-538 76693-540 76693-542
ɑ-Methylene-γ-butyrolactone 95% stabilized
alpha-Methylene-gamma-butyrolactone

Precaución: for Research Use Only

Formula: C₅H₆O₂
MW: 98.10 g/mol
MDL Number: MFCD00005407
CAS Number: 547-65-9
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR