cis-4-Cyclopentene-1,3-diol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
cis-Cyclopent-4-ene-1,3-diol
A307665-1G
A307665-250MG
76693-530EA
352.22
USD
76693-530
76693-532
cis-4-Cyclopentene-1,3-diol 98%
cis-4-Cyclopentene-1,3-diol
Precaución: for Research Use Only
Formula:
C₅H₈O₂ MW: 100.116 g/mol |
MDL Number:
MFCD00077703 CAS Number: 29783-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...