(S)-4-(4-((4-Chlorophenyl)(pyridin-2-yl)methoxy)piperidin-1-yl)butanoic acid compound with benzenesulfonic acid (1:1) 98%

Supplier: Ambeed
Warning

Synonyms: Bepotastine besilate

A307253-100MG A307253-10MG A307253-1MG A307253-250MG A307253-50MG A307253-1G A307253-5G
76693-520EA 35.01 USD
76693-520 76693-522 76693-524 76693-526 76693-528 77626-542 77626-544
(S)-4-(4-((4-Chlorophenyl)(pyridin-2-yl)methoxy)piperidin-1-yl)butanoic acid compound with benzenesulfonic acid (1:1) 98%
(S)-4-(4-((4-Chlorophenyl)(pyridin-2-yl)methoxy)piperidin-1-yl)butanoic acid compound with benzenesulfonic acid (1:1)

Precaución: for Research Use Only

Formula: C₂₇H₃₁ClN₂O₆S
MW: 547.06 g/mol
MDL Number: MFCD01938491
CAS Number: 190786-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR