Bromocyclohexane 98%

Supplier: Ambeed
Danger

A306392-1000G A306392-100G A306392-10G A306392-25G A306392-500G
76693-500EA 49 USD
76693-500 76693-502 76693-504 76693-506 76693-508
Bromocyclohexane 98%
Bromocyclohexane

Precaución: for Research Use Only

Formula: C₆H₁₁Br
MDL Number: MFCD00003819
CAS Number: 108-85-0
UN: 3082
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR