Calix[4]arene 98%

Supplier: Ambeed
Warning

Synonyms: 1,3,5,7(1,3)-Tetrabenzenacyclooctaphane-12,32,52,72-tetraol

A306389-1G A306389-5G
76693-496EA 51.99 USD
76693-496 76693-498
Calix[4]arene 98%
Calix[4]arene

Precaución: for Research Use Only

Formula: C₂₈H₂₄O₄
MW: 424.5 g/mol
MDL Number: MFCD00233673
CAS Number: 74568-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR