(4-Fluorophenoxy)acetic acid 98%

Supplier: Ambeed
Warning

A305533-100G A305533-5G A305533-25G A305533-500G
76693-492EA 52 USD
76693-492 77593-832 77601-380 77877-978
(4-Fluorophenoxy)acetic acid 98%
(4-Fluorophenoxy)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₄F)OCH₂COOH
MDL Number: MFCD00004304
CAS Number: 405-79-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR