(S)-2-Amino-4-phenylbutanoic acid hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: H-β-HoPhe-OH·HCl

A305476-1G A305476-5G
76693-488EA 21.22 USD
76693-488 76693-490
(S)-2-Amino-4-phenylbutanoic acid hydrochloride 98%
(S)-2-Amino-4-phenylbutanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₄ClNO₂
MW: 215.67 g/mol
MDL Number: MFCD03427064
CAS Number: 105382-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR