cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 98%

Supplier: Ambeed
Warning

Synonyms: (OC-6-22)-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate

A305416-100MG A305416-1G A305416-250MG
76693-482EA 31.83 USD
76693-482 76693-484 76693-486
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate 98%
cis-Bis(2,2'-bipyridine)dichlororuthenium(II) hydrate

Precaución: for Research Use Only

Formula: C₂₀H₁₈Cl₂N₄ORu
MW: 502.361 g/mol
MDL Number: MFCD00150005
CAS Number: 98014-14-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR