Carboplatin 99%

Supplier: Ambeed
Warning

Synonyms: cis-Diammine(1,1-cyclobutanedicarboxylato)platinum(II), cis-Diamine(1,1-cyclobutanedicarboxylato)platinum(II), 1,1-Cyclobutanedicarboxylatodiammineplatinum(II)

A305076-100MG A305076-1G A305076-1MG A305076-250MG A305076-5G
76693-464EA 27.59 USD
76693-464 76693-466 76693-468 76693-470 76693-472
Carboplatin 99%
Carboplatin

Precaución: for Research Use Only

Formula: C₆H₁₂N₂O₄Pt
MW: 371.26 g/mol
MDL Number: MFCD00070464
CAS Number: 41575-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR