6-Fluoro-4-quinolinol 98%

Supplier: Ambeed
Warning

A303255-1G A303255-250MG A303255-5G A303255-25G
76693-434EA 23 USD
76693-434 76693-436 76693-438 77877-973
6-Fluoro-4-quinolinol 98%
6-Fluoro-4-quinolinol

Precaución: for Research Use Only

Formula: C₉H₆FNO
MDL Number: MFCD00269610
CAS Number: 391-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR