2-(3,4-Dichlorophenyl)-1-(2-(pyrrolidin-1-ylmethyl)piperidin-1-yl)ethanone hydrochloride 95+%

Supplier: Ambeed
Warning

Synonyms: BRL 52537 hydrochloride

A300626-1MG A300626-5MG
76693-404EA 12.74 USD
76693-404 76693-406
2-(3,4-Dichlorophenyl)-1-(2-(pyrrolidin-1-ylmethyl)piperidin-1-yl)ethanone hydrochloride 95+%
2-(3,4-Dichlorophenyl)-1-(2-(pyrrolidin-1-ylmethyl)piperidin-1-yl)ethanone hydrochloride

Precaución: for Research Use Only

Formula: C₁₈H₂₅Cl₃N₂O
MW: 391.76 g/mol
MDL Number: MFCD00672679
CAS Number: 112282-24-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR