3-Ethynylaniline 98%

Supplier: Ambeed
Danger

Synonyms: 1-Amino-3-ethynylbenzene, (3-Aminophenyl)acetylene, (3-Aminophenyl)ethyne, 1-Ethynyl-3-aminobenzene

A300620-10G A300620-25G A300620-5G
76693-398EA 20 USD
76693-398 76693-400 76693-402
3-Ethynylaniline 98%
3-Ethynylaniline

Precaución: for Research Use Only

Formula: C₈H₇N
MDL Number: MFCD00014779
CAS Number: 54060-30-9
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR