Methyl-3-amino-2-methylbenzoate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-2-methylbenzoate, 3-Amino-o-toluic acid Methyl Ester, 3-Amino-2-methylbenzoic acid Methyl Ester

A298337-100G A298337-1G A298337-25G A298337-500G A298337-5G
76693-362EA 95.49 USD
76693-362 76693-364 76693-366 76693-368 76693-370
Methyl-3-amino-2-methylbenzoate 98%
Methyl 3-amino-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00211328
CAS Number: 18583-89-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR