4-(4-Aminophenyl)butanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4-(4-Aminophenyl)butyric acid

A297866-250MG A297866-10G A297866-1G A297866-25G
76693-350EA 9.55 USD
76693-350 77601-332 77626-522 77626-524
4-(4-Aminophenyl)butanoic acid 95%
4-(4-Aminophenyl)butanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂
MW: 179.2200 g/mol
MDL Number: MFCD00075052
CAS Number: 15118-60-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR