Boc-Piz HCl 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl piperazine-1-carboxylate hydrochloride

A295373-100G A295373-10G A295373-25G A295373-500G
76693-320EA 40.32 USD
76693-320 76693-322 76693-324 76693-326
Boc-Piz HCl 95%
Boc-Piz HCl

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₂·HCl
MW: 222.72 g/mol
MDL Number: MFCD00673776
CAS Number: 76535-74-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR