4,4'-(Phenylphosphoryl)dibenzoic acid 95%

Supplier: Ambeed
Warning

A294726-10G A294726-1G A294726-5G
76693-304EA 337.37 USD
76693-304 76693-306 76693-308
4,4'-(Phenylphosphoryl)dibenzoic acid 95%
4,4'-(Phenylphosphoryl)dibenzoic acid

Precaución: for Research Use Only

Formula: C₂₀H₁₅O₅P
MW: 366.30 g/mol
MDL Number: MFCD02093038
CAS Number: 803-19-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR