β-L-Adenosine 95%

Supplier: Ambeed
Warning

Synonyms: L-Adenosine, (2S,3S,4R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol

A293953-100MG A293953-250MG
76693-288EA 35.01 USD
76693-288 76693-290
β-L-Adenosine 95%
β-L-Adenosine

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O₄
MW: 267.2413 g/mol
MDL Number: MFCD01075784
CAS Number: 3080-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR