Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 95%

Supplier: Ambeed
Warning

Synonyms: Bis(2,2,2-trifluoroethyl)phosphonoacetic Acid Methyl Ester, Methyl [Bis(2,2,2-trifluoroethoxy)phosphinyl]acetate, Methyl Bis(2,2,2-trifluoroethyl)phosphonoacetate, Methyl O,O'-bis(2,2,2-trifluoroethyl)phosphonoacetate, Methyl 2-(bis(2,2,2-trifluoroethoxy)phosphoryl)acetate, Bis(2,2,2-trifluoroethyl) [(methoxycarbonyl)methyl]phosphonate

A293318-10G A293318-1G A293318-25G A293318-5G A293318-100G
76693-264EA 143.23 USD
76693-264 76693-266 76693-268 76693-270 77626-504
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate 95%
Bis(2,2,2-trifluoroethyl) (methoxycarbonylmethyl)phosphonate

Precaución: for Research Use Only

Formula: C₇H₉F₆O₅P
MW: 318.11 g/mol
MDL Number: MFCD00009901
CAS Number: 88738-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR