5,6,7,8-Tetrahydro-1-naphthol 98%

Supplier: Ambeed
Warning

A292753-100G A292753-1G A292753-250MG A292753-25G A292753-5G
76693-254EA 293.87 USD
76693-254 76693-256 76693-258 76693-260 76693-262
5,6,7,8-Tetrahydro-1-naphthol 98%
5,6,7,8-Tetrahydro-1-naphthol

Precaución: for Research Use Only

Formula: C₁₀H₁₂O
MW: 148.2 g/mol
MDL Number: MFCD00001734
CAS Number: 529-35-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR