5-Chloro-2-methylanisole 97%

Supplier: Ambeed
Danger

Synonyms: 4-Chloro-2-methoxy-1-methylbenzene

A291720-1G A291720-5G
76693-244EA 21.22 USD
76693-244 76693-246
5-Chloro-2-methylanisole 97%
5-Chloro-2-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉ClO
MW: 156.6095 g/mol
MDL Number: MFCD07368289
CAS Number: 40794-04-5
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR