(R)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate 95%

Supplier: Ambeed
Warning

A291135-250MG A291135-5G A291135-1G A291135-25G
76693-226EA 32 USD
76693-226 76693-228 77593-780 77626-502
(R)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate 95%
(R)-tert-Butyl (pyrrolidin-2-ylmethyl)carbamate

Precaución: for Research Use Only

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD03839886
CAS Number: 719999-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR