D-alpha-Hydroxyglutaric Acid Disodium 95%
Supplier: Ambeed
|
Warning
|
A289466-100MG
A289466-10G
A289466-10MG
A289466-1G
A289466-1MG
A289466-250MG
A289466-25MG
A289466-50MG
A289466-5G
76693-204EA
40
USD
76693-204
76693-206
76693-208
76693-210
76693-212
76693-214
76693-216
76693-218
76693-220
D-alpha-Hydroxyglutaric Acid Disodium 95%
D-alpha-Hydroxyglutaric Acid Disodium
Precaución: for Research Use Only
Formula:
C₅H₆Na₂O₅ |
MDL Number:
MFCD00069573 CAS Number: 103404-90-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...