2-Iodocumene 95%

Supplier: Ambeed
Warning

Synonyms: 2-Iodoisopropylbenzene, 1-Iodo-2-isopropylbenzene, 2-(2-Iodophenyl) propane

A289257-100G A289257-10G A289257-1G A289257-25G A289257-5G
76693-194EA 90.18 USD
76693-194 76693-196 76693-198 76693-200 76693-202
2-Iodocumene 95%
2-Iodocumene

Precaución: for Research Use Only

Formula: C₉H₁₁I
MW: 246.09 g/mol
MDL Number: MFCD00013704
CAS Number: 19099-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR