Diaveridine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Diaveridine HCl

A287796-1MG A287796-25MG A287796-50MG A287796-5MG
76693-174EA 8 USD
76693-174 76693-176 76693-178 76693-180
Diaveridine hydrochloride 98%
Diaveridine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₇ClN₄O₂
MW: 296.75 g/mol
MDL Number: MFCD07773087
CAS Number: 2507-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR